Masterflex® Fitting, Polyethylene, Straight, Thread Reducer, 3/8" NPT(M) x 1/2" NPT(M); 10/PK
Catalog # MFLX30623-52
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/8" NPT(M) x 1/2" NPT(M)
- Constructed of durable polyethylene
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
- Tipo di connessione:Threaded Reducer
- Flow pattern:Straight
- Thread size NPT(M) (port 1):⅜"
- Thread size NPT(M) (port 2):½"
- Descrizione:Fitting, Polyethylene, Straight, Thread Reducer, ⅜" NPT(M) x ½" NPT(M); 10/PK
- Seq:17
- Environmentally Preferable:
- Modello:30623-52
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:HDPE
Frequently Bought Together