Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 3/8" NPT(M); 10/PK
Codice catalogo MFLX30621-57
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/4" ID x 3/8" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/4"
- Tipo di connessione:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):3/8"
- Descrizione:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 3/8" NPT(M); 10/PK
- Seq:25
- Environmentally Preferable:
- Modello:30621-57
- No. of ports:2
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:Nylon
Frequently Bought Together