Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/2" ID x 3/4" NPT(M); 10/PK
Catalog # MFLX30621-21
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):1/2"
- Tipo di connessione:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):3/4"
- Descrizione:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/2" ID x 3/4" NPT(M); 10/PK
- Seq:14
- Environmentally Preferable:
- Modello:30621-21
- No. of ports:2
- Sterilizzazione:Ethylene oxide (ETO) only
- Conf:10
- Fitting material:Nylon
Frequently Bought Together