Barbed Straight Connector, 316SS, 5/16" ID
Codice catalogo MFLX30620-02
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):5/16"
- Dimensioni del raccordo/Ø int. del tubo (porta 2):5/16"
- Tipo di connessione:Barbed Union
- Flow pattern:Straight
- Descrizione:Barbed Straight Connector, 316SS, 5/16" ID
- Seq:02
- Environmentally Preferable:
- Modello:30620-02
- Orifice dimensions:3,175 mm (0,125")
- Sterilizzazione:Autoclave, ethylene oxide (ETO), or gamma
- Conf:1
- Fitting material:Stainless Steel 316
Frequently Bought Together