Masterflex® Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 10/Pk
Codice catalogo MFLX30600-03
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
- Flow pattern:Four-Way
- Descrizione:Fitting, Polycarbonate, Four-Way Stopcock, Male Slip Luer, Non-Sterile; 10/Pk
- Environmentally Preferable:
- Materiali umidi:Polycarbonate
- Garanzia:Out of box
- Conf:10
- Inlet port:Female Luer
- Outlet port:Male Luer
Frequently Bought Together