Masterflex® Smart Gasket® Sampling Injection Tube for System, Stainless Steel; 1/EA
Codice catalogo MFLX30545-99
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Optimize performance of your Masterflex® Smart Gasket® Sanitary Gasket system.
- Choose from a wide range of replacement parts and accessories
Smart Gaskets allow you to measure temperature, take samples, or dose chemicals in your process line. Selected accessories include thermocouple probes and wire, port plugs for the Smart Gasket, and a sampling tube.
- Tipo di collegamento:Sanitary
- Descrizione:Sampling Injection Tube for System, Stainless Steel; 1/EA
- Seq:02
- Environmentally Preferable:
- Conf:1
- Fitting material:Stainless Steel
- Garanzia:1 year
Frequently Bought Together