PendoTECH® Single-Use Pressure Sensor Accessories
Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
Codice catalogo MFLX19406-14
CE Compliant
Nome del fornitore: Avantor Fluid Handling
PendoTECH® Single-Use Pressure Sensor Accessories
Codice catalogo MFLX19406-14
Nome del fornitore: Avantor Fluid Handling
CE Compliant
Vai a
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Descrizione:Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
- Cat. No.:MFLX19406-14
- Lunghezza:213,36 cm (84")
- Garanzia:2 years