Masterflex® L/S® Peristaltic Pump Drive Replacement Brushes; 2/PK
Codice catalogo MFLX07533-01
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Quickly and easily replace components of your Masterflex system.
- Choose replacement brushes and caps based on your Masterflex L/S drive
- Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
- Descrizione:L/S® Replacement Brushes, 2/PK
- Environmentally Preferable:
- Conf:2
Frequently Bought Together