Masterflex® L/S® Standard Pump Head, Replacement Rotor Assembly, High-Performance Precision Tubing Sizes, CRS Rotor
Codice catalogo MFLX07015-80
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07015-20 and -42; 07024-20 and -42
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Descrizione:Standard Pump Head, Replacement Rotor Assembly, High-Performance Precision Tubing Sizes, CRS Rotor; 1/Ea
- Seq:09
- Environmentally Preferable:
- Modello:07015-80
- Open-head sensor:No
- Tubing sizes accepted:L/S 15, 24, 35, 36
- Garanzia:2 years
- Conf:1
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together