Masterflex® L/S® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14
Catalog # MFLX07014-81
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07014-00, -20, -70, and -72
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Descrizione:Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, L/S® 14 Tubing
- Seq:06
- Environmentally Preferable:
- Modello:07014-81
- Open-head sensor:No
- Tubing sizes accepted:L/S 14
- Garanzia:2 years
- Conf:1
- Housing material:PC
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together