Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, three pump heads, stainless steel; 2/set, 2 years warranty, Acciaio inossidabile, Masterflex L/S® Easy-Load® Pump Head, L/S
Codice catalogo MFLX07013-08
Nome del fornitore: Avantor Fluid Handling
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Codice catalogo MFLX07013-08
Nome del fornitore: Avantor Fluid Handling
Vai a
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Certificates
Cerca per numero di lottoSpecifiche
- Descrizione:L/S® Pump head mounting hardware, Standard and Easy-Load®, three pump heads, stainless steel; 2/set, 2 years warranty
- Materiale:Acciaio inossidabile
- Seq:03
- Compatibile con:Masterflex L/S® Easy-Load® Pump Head
- Pump series:L/S
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.