Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Tripod Base for Multi-Tube Frames, Acrylic
Codice catalogo MFLX03226-50
Nome del fornitore: Avantor Fluid Handling
Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Codice catalogo MFLX03226-50
Nome del fornitore: Avantor Fluid Handling
Vai a
Product Details & Documents
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
Specifiche
- Descrizione:Tripod Base for Multi-Tube Frames
- Seq:05
- Housing material:Acrylic
- Cat. No.:MFLX03226-50
- Modello:03226-50
- Garanzia:1 year
- Larghezza:1755"