0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
CYNOMOLGUS MONKEY IL-1 BETA (IL-1F2) (YEAST-DERIVED RECOMBINANT PROTEIN) 1 * 25 µG
KINFRP1597Y-025
:
Kingfisher Biotech
CYNOMOLGUS MONKEY IL-1 BETA (IL-1F2) (YEAST-DERIVED RECOMBINANT PROTEIN) 1 * 25 µG
KINFRP1597Y-025
:
Kingfisher Biotech
Frequently Bought Together
VWR®, Contenitori per campioni dritti con tappo a vite
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Provette e strip per PCR, 0,2 ml
: VWR Collection
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
VWR®, Divaricatori, a L, monouso
: VWR Collection
Provette per microcentrifuga Eppendorf® Protein LoBind®
: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF