[EN]METHYLCYCLOHEXANE REAGENT 1 * 500 mL
Katalognr. SPCMM1249-500ML
Anbieter: Spectrum Chemical
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Metanolo ≥99.9% (by GC), HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Toluene ≥99.8%, SupraSolv® for GC-MS
Anbieter: Merck
Glicerina 85% Ph. Eur.
Anbieter: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
Anbieter: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals
Tetraidrofurano ≥99.9% non stabilizzato, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals