Coupling body plugs
Coupling body plug, with class VI silicone O-ring, MPC series, Policarbonato
Catalog # CPCCMPC30003MB25
Supplier: CPC COLDER
Jump to:
Product Details & Documents
Class VI coupling body plug with Class VI silicone O-ring.
Documents
Specifications
- Descrizione:Coupling body plug, with class VI silicone O-ring, MPC series
- Materiale:Policarbonato
- Seq:01