[EN]HOSE BARB 1/2 POLYS. COUPLING SET 1 * 10 pezzi
Catalog # CPCCHFCD39SET8HC
Supplier: CPC COLDER
Frequently Bought Together
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Plastic Assorted Fitting Kits, Avantor®
Supplier: Avantor Fluid Handling
SaniSure® Single-Use Sanitary Clamp Accessories
Supplier: Avantor Fluid Handling
[EN]00125KI-HOSE BARB IN-LINE 3/8IN 25PK 1 * 25 pezzi
Supplier: Avantor Fluid Handling
[EN]COUPL. BODY PLUG POLYS. USP CLASS O- 1 * 10 pezzi
Supplier: CPC COLDER
Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Male Luer to Barb Fitting, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Supplier: Avantor Fluid Handling
[EN]1/8 X 3/32 COMMERCIAL GRADE 1 * 1 pezzi
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hosebarb to NPT(M), Elbow, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Certified Sanitary Adapters, ADCF Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, Nylon, Avantor®
Supplier: Avantor Fluid Handling
[EN]FITTING 1/4 BARBX1/4 MALE NPT 1 * 1 pezzi
Supplier: BIOTAGE
Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Tuf-Steel® Sanitary Tri-Clamp® Gaskets, Avantor®
Supplier: Avantor Fluid Handling
[EN]ELBOW PFA 3/8''FL X 1/4''MNPT 1 * 1 pezzi
Supplier: Avantor Fluid Handling
Fitting, S/S, 1/4 B3 x 3-SS 1 * 1 items
Supplier: BIOTAGE