[EN]BINLINER 34INX40INX2MIL RED BIOH.BUL 1 * 50 pezzi
Catalog # MICRBL2-3440REBB
Supplier: MICRONOVA
[EN]BINLINER 34INX40INX2MIL RED BIOH.BUL 1 * 50 pezzi
Catalog # MICRBL2-3440REBB
Supplier: MICRONOVA
Frequently Bought Together
QuickTask™ flat mops
Supplier: CONTEC
[EN]BOOT JALAS 9975 EXALTER EU34/2 1 * 1 PAIO
Supplier: EJENDALS
Safety glasses, SecureFit™ 500 series
Supplier: 3M
RAIN TROUSERS BLACK F441 3XL 1 * 1 pezzi
Supplier: Portwest
Accessories for cleaning cart
Supplier: Rubbermaid Commercial Products
[EN]HOSE CLAMP 160MM PLASTIC COATED 1 * 1 pezzi
Supplier: HAMMACHER KARL
Steckschlüssel-Sätze WERA 9720 8000 A Zyklop Knarre 1 4 Set 1 1 * 1 SET
Supplier: MARKET SOURCE PART PROCESS
[EN]BOOT JALAS 9975 EXALTER EU44/10 1 * 1 PAIO
Supplier: EJENDALS
Modular deck system, PIG®
Supplier: NEW PIG
Brady® Vinyl Labels for BradyJet™ J7300 Colour Label Printer
Supplier: Brady
CleanMax® Clean Manufactured Coveralls with Attached Hood and Boots
Supplier: LAKELAND CLEANMAX APPAREL