Orders LinkContactUsLinkComponent
Wash troughs
Wash troughs
Infra-red mains operated sensor four station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2000W×380D×1250H mm, 1990×123×283 mm
Codice catalogo TEKMWTN040EXSENPED
Nome del fornitore:   TEKNOMEK
Wash troughs
Codice catalogo TEKMWTN040EXSENPED
Nome del fornitore:  TEKNOMEK
Wash troughs
Wash troughs
Infra-red mains operated sensor four station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2000W×380D×1250H mm, 1990×123×283 mm
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
Informazioni per l'ordine: Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
Specifiche
  • Descrizione:
    Infra-red mains operated sensor four station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2000W×380D×1250H mm
  • Seq:
    23
  • Interno LxPxA:
    1990×123×283 mm