Orders LinkContactUsLinkComponent
Wash troughs
Wash troughs
Infra-red mains operated sensor two station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 1010W×380D×1250H mm, 1000×123×283 mm
N° de catalogue TEKMWTN020EXSENPED
Fournisseur:   TEKNOMEK
Wash troughs
N° de catalogue TEKMWTN020EXSENPED
Fournisseur:  TEKNOMEK
Wash troughs
Wash troughs
Infra-red mains operated sensor two station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 1010W×380D×1250H mm, 1000×123×283 mm
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
Informations de commande: Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
Spécifications
  • Descrizione:
    Infra-red mains operated sensor two station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 1010W×380D×1250H mm
  • Seq:
    12
  • Interno LxPxA:
    1000×123×283 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.