Orders LinkContactUsLinkComponent
Wash troughs
Wash troughs
Infra-red mains operated sensor one station wash trough, wall mounted, fully welded 304-grade stainless steel, 410W×370D×525H mm, 400×123×283 mm
Codice catalogo TEKMWTN010EXSEN
Nome del fornitore:   TEKNOMEK
Wash troughs
Codice catalogo TEKMWTN010EXSEN
Nome del fornitore:  TEKNOMEK
Wash troughs
Wash troughs
Infra-red mains operated sensor one station wash trough, wall mounted, fully welded 304-grade stainless steel, 410W×370D×525H mm, 400×123×283 mm
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
 : Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
Specifiche
  • Descrizione:
    Infra-red mains operated sensor one station wash trough, wall mounted, fully welded 304-grade stainless steel, 410W×370D×525H mm
  • Seq:
    03
  • Interno LxPxA:
    400×123×283 mm