Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Interior and surface cleaners, SURE®

Interior and surface cleaners, SURE®

Interior and surface cleaner, 6×1 L
Codice catalogo 115-2960
   Materiali sostenibili|Più sicuro dal punto di vista della salute umana e dell'ambiente|Imballi sostenibili
Nome del fornitore:   Diversey
Interior and surface cleaners, SURE®
Codice catalogo 115-2960
Nome del fornitore:  Diversey
   Materiali sostenibili|Più sicuro dal punto di vista della salute umana e dell'ambiente|Imballi sostenibili
Interior and surface cleaners, SURE®
Interior and surface cleaners, SURE®

Interior and surface cleaner, 6×1 L

Price
SFr. 38.90
Log in per disponibilità
1 * 6 Fl
Product Details & Documents

Concentrated all-purpose detergent for cleaning of hard surfaces, such as floors, walls, doors, ceilings and general equipment. Can also be used in food premises.

  • Suitable for cleaning of all water resistant hard surfaces, including aluminium and other soft metals
  • Effective in all water conditions
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for user
  • Concentrated formulation gives excellent economy of use
  • Fast acting, fast drying

SURE® interior and surface cleaner is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.

Certifications: EU-Ecolabel (EU:NL/020/045).
The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.

Specifiche
  • Descrizione:
    Interior and surface cleaner, 6×1 L
  • Seq:
    01
  • Cat. No.:
    115-2960
  • Environmentally Preferable:
    Materiali sostenibili|Più sicuro dal punto di vista della salute umana e dell'ambiente|Imballi sostenibili
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.