Orders LinkContactUsLinkComponent
VWR®, Vial Racks
VWR®, Vial Racks
Vial rack for 4 ml vials, transparent, acryl, 15,2 mm, 40, 175,8×115,5×20 mm
  548-0192A
VWR®, Vial Racks
  548-0192A
VWR®, Vial Racks
VWR®, Vial Racks
Vial rack for 4 ml vials, transparent, acryl, 15,2 mm, 40, 175,8×115,5×20 mm
107.00 SFr.
1 * 1 ST
Vial Racks are ideal for storage and transportation of vials.
  • Safe storage of vials
  • Stable construction
  • Easy handling and transportation
These vial racks allow the vials to stand securely on the laboratory bench. The empty racks are stackable and therefore space-saving.
  • Ø foro:
    15,2 mm
  • N. di fori:
    40
  • Descrizione:
    Vial rack for 4 ml vials, transparent, acryl
  • Seq:
    01
  • L×P×A:
    175,8×115,5×20 mm