Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Snap Cap Vials

VWR®, Snap Cap Vials

Vial kits, 25,00 ml, Flat bottom, 30×50 mm, trasparente, Yes, ND28, PE, Colour-21390374|Colour-21390348
Codice catalogo 548-0649A
Nome del fornitore:   VWR Collection
VWR®, Snap Cap Vials
Codice catalogo 548-0649A
Nome del fornitore:  VWR Collection
VWR®, Snap Cap Vials
VWR®, Snap Cap Vials

Vial kits, 25,00 ml, Flat bottom, 30×50 mm, trasparente, Yes, ND28, PE, Colour-21390374|Colour-21390348

Price
SFr. 97.90
Log in per disponibilità
1 * 250 ST
Product Details & Documents

Snap cap vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They should only be used for storage of solids and powders.

  • No liners needed in the cap
  • Good handling with gloves
  • Quick and easy open/close
  • Easy filling

Snap cap vials constructed from type I borosilicate glass with low metal content. Vials are available in clear version with a flat bottom without a cap or as a kit with a PE cap. The usage of these vials allows the samples to be stored for a long time (long-term storage applications).

Specifiche
  • Tappo incluso:
    Yes
  • Materiale tappo:
    PE
  • Dimensione tappo:
    ND28
  • Ø×A:
    30×50 mm
  • Capacità:
    25,00 ml
  • Colore:
    trasparente
  • Descrizione:
    Vial kits
  • Seq:
    11
  • Colore tappo:
    Colour-21390374|Colour-21390348
  • Fondo:
    Flat bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.