Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Snap Cap Vials

VWR®, Snap Cap Vials

Vials, 5,00 ml, Flat bottom, 20×40 mm, Borosilicate glass 5.1, trasparente, No, ND18
Codice catalogo 548-0141A
Nome del fornitore:   VWR Collection
VWR®, Snap Cap Vials
Codice catalogo 548-0141A
Nome del fornitore:  VWR Collection
VWR®, Snap Cap Vials
VWR®, Snap Cap Vials

Vials, 5,00 ml, Flat bottom, 20×40 mm, Borosilicate glass 5.1, trasparente, No, ND18

Price
SFr. 564.00
Log in per disponibilità
1 * 1.000 ST
Product Details & Documents

Snap cap vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They should only be used for storage of solids and powders.

  • No liners needed in the cap
  • Good handling with gloves
  • Quick and easy open/close
  • Easy filling

Snap cap vials constructed from type I borosilicate glass with low metal content. Vials are available in clear version with a flat bottom without a cap or as a kit with a PE cap. The usage of these vials allows the samples to be stored for a long time (long-term storage applications).

Specifiche
  • Tappo incluso:
    No
  • Dimensione tappo:
    ND18
  • Ø×A:
    20×40 mm
  • Capacità:
    5,00 ml
  • Colore:
    trasparente
  • Descrizione:
    Vials
  • Materiale:
    Borosilicate glass 5.1
  • Seq:
    01
  • Fondo:
    Flat bottom