System waste caps, b.safe
Waste caps, filling below surface level, S 55, 1× GL 25 (m) hose connector, 1× GL15 (f) aeration connector, PPS
Codice catalogo BOHLR1321-55
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
b.safe waste tubes can be connected to the canister by means of these b.safe system waste caps and solvents can be collected safely. Solvent vapours are held back by the exhaust filter with activated carbon or can be led to the hood by means of a venting hose.
- Made of high-performance plastics
- Thermally and chemically stable
- Good grip for easy assembly, safe working and improved processes
Screw cap made of PPS, free movable insert made of conductible PE-EX with 1 connector GL 25 suitable for b.safe Waste Tubes and 1 connector GL 14 for fixing the b.safe Venting Hose or a b.safe Exhaust Filter.
Design and scope of delivery according to table.
Specifiche
- Filettatura:S 55
- Descrizione:Waste caps, filling below surface level
- Materiale:PPS
- Seq:03
- Connettore:1× GL 25 (m) hose connector, 1× GL15 (f) aeration connector