ContactUsLinkComponent
HCoV-229E Spike Protein, His Tag (SPN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 1115 (Accession # P15423-1 (TI 871-872 PP)).The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trim 1 * 500 µG
Codice catalogo ACRBSPN-H52H3500UG
Nome del fornitore:  ACRO Biosystems
HCoV-229E Spike Protein, His Tag (SPN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 1115 (Accession # P15423-1 (TI 871-872 PP)).The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trim 1 * 500 µG
Codice catalogo ACRBSPN-H52H3500UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together