HCoV-HKU1 (isolate N5) Spike Trimer, His Tag (SPN-H52H5) is the ectodomain of HCoV-HKU1 (isolate N5) spike protein which contains AA Ala 13 - Asn 1276 (Accession # Q0ZME7-1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 1 * 500 µG
Codice catalogo ACRBSPN-H52H5500UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together