ContactUsLinkComponent
FITC-Labeled Human CD37, Fc Tag (CD7-HF26x) is expressed from human 293 cells (HEK293). It contains AA Arg 112 - Asn 241 (Accession # P11049-1). 1 * 25 µG
Catalog # ACRBCD7-HF26X-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CD37, Fc Tag (CD7-HF26x) is expressed from human 293 cells (HEK293). It contains AA Arg 112 - Asn 241 (Accession # P11049-1). 1 * 25 µG
Catalog # ACRBCD7-HF26X-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together