ContactUsLinkComponent
FITC-Labeled Human CD30, His Tag (CD0-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). 1 * 200 µG
N° de catalogue ACRBCD0-HF2H4200UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human CD30, His Tag (CD0-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2). 1 * 200 µG
N° de catalogue ACRBCD0-HF2H4200UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together