ContactUsLinkComponent
FITC-Labeled Human IL-3 R alpha, Fc Tag (ILA-HF253) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305  (Ac cession # P26951-1). 1 * 25 µG
Catalog # ACRBILA-HF253-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human IL-3 R alpha, Fc Tag (ILA-HF253) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305  (Ac cession # P26951-1). 1 * 25 µG
Catalog # ACRBILA-HF253-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together