FITC-Labeled Human IL-3 R alpha, Fc Tag (ILA-HF253) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Ac cession # P26951-1). 1 * 200 µG
Katalognr. ACRBILA-HF253200UG
Anbieter: ACROBIOSYSTEMS
Frequently Bought Together