ContactUsLinkComponent
Human CD79B, His Tag (CDB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1). 1 * 100 µG
Codice catalogo ACRBCDB-H52H3100UG
Nome del fornitore:  ACRO Biosystems
Human CD79B, His Tag (CDB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Accession # P40259-1). 1 * 100 µG
Codice catalogo ACRBCDB-H52H3100UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together