ContactUsLinkComponent
Human CCR5 Full Length Protein-VLP (CC5-H52P3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Leu 352 (Accession # P51681-1). 1 * 100 µG
Codice catalogo ACRBCC5-H52P3100UG
Nome del fornitore:  ACROBIOSYSTEMS
Human CCR5 Full Length Protein-VLP (CC5-H52P3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Leu 352 (Accession # P51681-1). 1 * 100 µG
Codice catalogo ACRBCC5-H52P3100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together