ContactUsLinkComponent
Human CCR8 Full Length Protein-VLP (CC8-H52P4) is expressed from human 293 cells (HEK293). It contains AA Asp 2 - Leu 355 (Accession # P51685-1). 1 * 100 µG
Codice catalogo ACRBCC8-H52P4100UG
Nome del fornitore:  ACROBIOSYSTEMS
Human CCR8 Full Length Protein-VLP (CC8-H52P4) is expressed from human 293 cells (HEK293). It contains AA Asp 2 - Leu 355 (Accession # P51685-1). 1 * 100 µG
Codice catalogo ACRBCC8-H52P4100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together