ContactUsLinkComponent
Human Nucleolin, Fc Tag (NUL-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accession # P19338-1 ). 1 * 100 µG
Codice catalogo ACRBNUL-H5253100UG
Nome del fornitore:  ACROBIOSYSTEMS
Human Nucleolin, Fc Tag (NUL-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accession # P19338-1 ). 1 * 100 µG
Codice catalogo ACRBNUL-H5253100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together