Human Latent TGF-beta 3 / Latent TGFB3, Tag Free (TG3-H5213) is expressed from human 293 cells (HEK293). It contains AA Ala 301 - S er 412 (Accession # P10600-1). 1 * 10 µG
Codice catalogo ACRBTG3-H5213-10UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together