ContactUsLinkComponent
FITC-Labeled Human B7-H3 (4Ig), Fc Tag (B7B-HF2F4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Acce ssion # Q5ZPR3-1). It is the FITC labeled form of Human B7-H3 (4Ig), Fc Tag (Cat. No. B7B-H5258). 1 * 25 µG
Catalog # ACRBB7B-HF2F4-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human B7-H3 (4Ig), Fc Tag (B7B-HF2F4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Acce ssion # Q5ZPR3-1). It is the FITC labeled form of Human B7-H3 (4Ig), Fc Tag (Cat. No. B7B-H5258). 1 * 25 µG
Catalog # ACRBB7B-HF2F4-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together