Watch glasses
Watch glass, glass, with fused rim, 120 mm
Codice catalogo HECH40010012
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
- Good chemical restistance
- Available with or without hole
Certifications: DIN 12 341
Specifiche
- Ø:120 mm
- Descrizione:Watch glass, glass, with fused rim
- Seq:18