Cooling Blocks, Passive Cooler
Cooling block, passive cooler, PP/aluminium, 96 well plates
N° de catalogue BRND709513
Fournisseur: Brand
Aller à
Product Details & Documents
Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.
ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station
- Cools microcentrifuge tubes and PCR consumables
All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.
Documents
Cooling Blocks.pdf
Notes d'application
Manual Cooling Block.pdf
Manuel
TDS Liquid Handling Station.pdf
Fiche technique
Spécifications
- Descrizione:Cooling block, passive cooler, PP/aluminium
- Seq:04
- Per:96 well plates
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.