Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Cooling Blocks, Passive Cooler

Cooling block, passive cooler, 9 positions, PP/aluminium, 5 ml micro tubes
Catalog # BRND709512
Supplier:   Brand
Cooling Blocks, Passive Cooler
Catalog # BRND709512
Supplier:  Brand
Cooling Blocks, Passive Cooler
Cooling Blocks, Passive Cooler

Cooling block, passive cooler, 9 positions, PP/aluminium, 5 ml micro tubes

Price
SFr. 1’170.00
Log in for availability
1 * 1 ST
Product Details & Documents

Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.

  • ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station

  • Cools microcentrifuge tubes and PCR consumables

All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.

Specifications
  • Descrizione:
    Cooling block, passive cooler, 9 positions, PP/aluminium
  • Seq:
    03
  • Per:
    5 ml micro tubes