Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cooling Blocks, Passive Cooler

Cooling Blocks, Passive Cooler

Cooling block, passive cooler, 16 positions, PP/aluminium, 1,5 ml micro tubes
Codice catalogo BRND709511
Nome del fornitore:   Brand
Cooling Blocks, Passive Cooler
Codice catalogo BRND709511
Nome del fornitore:  Brand
Cooling Blocks, Passive Cooler
Cooling Blocks, Passive Cooler

Cooling block, passive cooler, 16 positions, PP/aluminium, 1,5 ml micro tubes

Price
SFr. 1’170.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.

  • ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station

  • Cools microcentrifuge tubes and PCR consumables

All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.

Specifiche
  • Descrizione:
    Cooling block, passive cooler, 16 positions, PP/aluminium
  • Seq:
    02
  • Per:
    1,5 ml micro tubes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.