BD Eclipse™ Needles
Needle, sterile, 21 G, 25 mm
Codice catalogo BDAM305894
Nome del fornitore: BD Medical
Vai a
Product Details & Documents
The BD Eclipse™ SmartSlip™ is a single use sterile needle with a safety mechanism that covers the needle point after use. In the activated position, the needle cover guards against accidental needlestick during normal handling and disposal of the used needle/syringe.
- The SmartSlip™ Technology is specially designed for safe assembling of BD Eclipse™ needles to Luer slip syringes
- Bevel oriented to the safety cover to allow low angle injections
- By assembling BD Eclipse™ needles to a luer slip syringe, an audible click indicates a safely assembled needle
- Safety can be activated immediately after injection
- Wide, textured finger pad for thumb or forefinger
- Single-handed with one finger activation of the safety shield, audible click indicates that the cover has safely engaged the needle
- Natural Rubber Latex is not part of the material formulation
- Sterile, single use
The SmartSlip™ Technology consists of an adaptor to enable connection of the BD Eclipse safety needle to a Luer slip.
Specifiche
- Gauge:21 G
- Descrizione:Needle, sterile
- Lunghezza:25 mm