ContactUsLinkComponent
Rottlerin, Chemical. CAS: 82-08-6. Formula: C30H28O8. MW: 516.5. Anticancer agent that can inhibit cell growth, induce apoptosis, au tophagy and cell cycle arrest, inhibit cell invasion and shows anti-angiogenic activity. OXPHOS inhibitor. Mitochondr 1 * 50 mg
  ADIPAGCN20526M050
Rottlerin, Chemical. CAS: 82-08-6. Formula: C30H28O8. MW: 516.5. Anticancer agent that can inhibit cell growth, induce apoptosis, au tophagy and cell cycle arrest, inhibit cell invasion and shows anti-angiogenic activity. OXPHOS inhibitor. Mitochondr 1 * 50 mg
  ADIPAGCN20526M050
Frequently Bought Together