Zero pressure weather caps
Zero pressure weather caps,914 mm height,152 mm nominal diameter,9,1 kg shipping weight
Codice catalogo LBCP4722200
Nome del fornitore: LABCONCO
Vai a
Product Details & Documents
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Specifiche
- Descrizione:Zero pressure weather caps,914 mm height,152 mm nominal diameter,9,1 kg shipping weight
- Seq:01