Orders LinkContactUsLinkComponent
Accessories for Stuart Temperature Controller SCT2
Accessories for Stuart Temperature Controller SCT2
Stuart® replacement temperature probe for temperature controller, stainless steel, temperature range 50...200 °C with ±0,5 °C accuracy
Codice catalogo STUA99955-86
Nome del fornitore:   Cole Parmer
Accessories for Stuart Temperature Controller SCT2
Codice catalogo STUA99955-86
Nome del fornitore:  Cole Parmer
Accessories for Stuart Temperature Controller SCT2
Accessories for Stuart Temperature Controller SCT2
Stuart® replacement temperature probe for temperature controller, stainless steel, temperature range 50...200 °C with ±0,5 °C accuracy
Price
271.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Descrizione:
    Stuart® replacement temperature probe for temperature controller, stainless steel, temperature range 50...200 °C with ±0,5 °C accuracy
  • Seq:
    04