Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Solfanilammide

Solfanilammide

Bottle, 100 g
Catalog # APOSBIMB1014-100G
Supplier:   Apollo Scientific
Solfanilammide
Catalog # APOSBIMB1014-100G
Supplier:  Apollo Scientific
CAS Number:  63-74-1
Solfanilammide

Bottle, 100 g

CAS Number:  63-74-1
Price
SFr. 29.50
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Aminobenzenesulphonamide
  • CAS Number:63-74-1
  • Formula:H₂NC₆H₄SO₂NH₂
  • MDL Number:MFCD00007939
  • Molecular Weight:172.21 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.