D-(-)-Lyxose
Bottle, 5 g
Catalog # APOSBIFS0109-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:1114-34-7
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00064362
- Molecular Weight:150.13 g/mol
- Melting Pt:108…115 °C
- Storage Temperature:Ambiente
- EINECS:214-212-8
Specifications
Specifications
- Tipo di conf.:Bottle
- Conf:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



