Fluorite Objectives for Revolve Microscope
Plan fluorite dry CC; 0,95 NA, 100×, 0,20 mm
Katalognr. ECHLJMAG1011
Anbieter: DISCOVER ECHO
Direkt zu
Product Details & Documents
Dokumente
ECHO Meet Revolve
Produktliteratur
Technische Daten
- Ingrandimento:100×
- Descrizione:Plan fluorite dry CC; 0,95 NA
- Seq:09
- Distanza di lavoro:0,20 mm