Rat Interferon gamma ELISPOT Set
Rat Interferon gamma ELISPOT set, 20×96 tests
N° de catalogue ABCAAB46590-20X96
Fournisseur: Abcam
Aller à
Product Details & Documents
Rat Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Rat Interferon gamma production in a single cell suspension.
Spécifications
Spécifications
- Descrizione:Rat Interferon gamma ELISPOT set
- Test per kit:20×96 tests
- Cat. No.:ABCAAB46590-20X96
- Forma:Uncoated
- Stato regolamentazione:RUO
- Tiipo del campione:Suspension cells
- Temperatura di conservazione:4 ℃
- Tipo di anticorpi:Interferon gamma
- Tempo di elaborazione:Multiple steps
- Reattività delle specie:Rat
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



