Human IL-1 beta ELISPOT Set
Human IL-1 beta ELISPOT set, 20×96 tests
ABCAAB46578-20X96
: Abcam
Human IL-1 beta ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-1 beta production in a single cell suspension.
- Descrizione:Human IL-1 beta ELISPOT set
- Test per kit:20×96 tests
- Cat. No.:ABCAAB46578-20X96
- Tipo di prodotto:Uncoated
- Simbolo dell'antigene:IL-1 beta
- Stato regolamentazione:RUO
- Tiipo del campione:Suspension cells
- Temperatura di conservazione:4° C
- Tempo di elaborazione:Multiple steps
- Reattività delle specie:Human
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



